Merck
Comparative genomic analysis of eutherian ribonuclease A genes.
Molecular genetics and genomics : MGG (2013-12-18)
Marko Premzl
ABSTRACT
The present study attempted to update comprehensive eutherian ribonuclease A gene data sets, using public eutherian genomic sequence data sets and new genomics and molecular evolution tests. Among 448 ribonuclease A potential coding sequences, the present analysis annotated 255 complete coding sequences. The most comprehensive data set of eutherian ribonuclease A genes first characterized 13 major gene clusters, 9 of which showed evidence of differential gene expansions. In addition, the present analysis described common predicted promoter regions of eutherian ribonuclease A genes. The present study also attempted to resolve discrepancies in descriptions of eutherian ribonuclease A genes. Thus, the integrated gene annotations, phylogenetic analysis and protein molecular evolution analysis proposed new classification and nomenclature of eutherian ribonuclease A genes, as new framework of future experiments.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 4×cryst., ~70 U/mg (acc. to Kunitz)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein