Merck
  • von Willebrand factor mutation promotes thrombocytopathy by inhibiting integrin αIIbβ3.
von Willebrand factor mutation promotes thrombocytopathy by inhibiting integrin αIIbβ3.
The Journal of clinical investigation (2013-11-26)
Caterina Casari, Eliane Berrou, Marilyne Lebret, Frédéric Adam, Alexandre Kauskot, Régis Bobe, Céline Desconclois, Edith Fressinaud, Olivier D Christophe, Peter J Lenting, Jean-Philippe Rosa, Cécile V Denis, Marijke Bryckaert
ABSTRACT
von Willebrand disease type 2B (vWD-type 2B) is characterized by gain-of-function mutations in von Willebrand factor (vWF) that enhance its binding to the glycoprotein Ib-IX-V complex on platelets. Patients with vWD-type 2B have a bleeding tendency that is linked to loss of vWF multimers and/or thrombocytopenia. In this study, we uncovered evidence that platelet dysfunction is a third possible mechanism for bleeding tendency. We found that platelet aggregation, secretion, and spreading were diminished due to inhibition of integrin αIIbβ3 in platelets from mice expressing a vWD-type 2B-associated vWF (vWF/p.V1316M), platelets from a patient with the same mutation, and control platelets pretreated with recombinant vWF/p.V1316M. Impaired platelet function coincided with reduced thrombus growth. Further, αIIbβ3 activation and activation of the small GTPase Rap1 were impaired by vWF/p.V1316M following exposure to platelet agonists (thrombin, ADP, or convulxin). Conversely, thrombin- or ADP-induced Ca2+ store release, which is required for αIIbβ3 activation, was normal, indicating that vWF/p.V1316M acts downstream of Ca2+ release and upstream of Rap1. We found normal Syk phosphorylation and PLCγ2 activation following collagen receptor signaling, further implying that vWF/p.V1316M acts directly on or downstream of Ca2+ release. These data indicate that the vWD-type 2B mutation p.V1316M is associated with severe thrombocytopathy, which likely contributes to the bleeding tendency in vWD-type 2B.
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type XIV, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type I, lyophilized powder, 10-50 units/mg protein