Merck
  • [Glucagon-like peptide-1 regulates lipometabolism by down-regulating adipose triglyceride lipase in 3T3-L1 adipocytes].
[Glucagon-like peptide-1 regulates lipometabolism by down-regulating adipose triglyceride lipase in 3T3-L1 adipocytes].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2013-10-23)
Huili Zhu, Zeping Weng, Chenli Lin, Jiwei Ma, Xueyun Zhong
ABSTRACT
To investigate the effect of glucagon-like peptide-1 (GLP-1) on glycolipid metabolism in 3T3-L1 adipocytes and explore the mechanism. 3T3-L1 adipocytes were treated with GLP-1, insulin, or both for 24 h, and Western blotting was used to analyze the expression levels of adipose triglyceride lipase (ATGL), glucose transporter type 4 (GLUT4), Akt1, Akt2 and phosphorylated Akt in the cells. Immunofluorescence was used to observe lipid content in 3T3-L1 adipocytes. Akt1 and Akt2 were not activated by insulin stimulation in 3T3-L1 adipocytes. Akt was phosphorylated by GLP-1 stimulation, which inhibited the expression of ATGL and increased the translocation of GLUT4 from the intracellular membranes to plasma membranes. These changes were more obvious under the synergistic effect of insulin in 3T3-L1 adipocytes. GLP-1 decreases lipolysis by inhibiting the expression of ATGL and improves insulin resistance by increasing the translocation of GLUT4 in 3T3-L1 adipocytes.
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells