Merck
  • The fate of tandemly duplicated genes assessed by the expression analysis of a group of Arabidopsis thaliana RING-H2 ubiquitin ligase genes of the ATL family.
The fate of tandemly duplicated genes assessed by the expression analysis of a group of Arabidopsis thaliana RING-H2 ubiquitin ligase genes of the ATL family.
Plant molecular biology (2013-10-19)
Victor Aguilar-Hernández, Plinio Guzmán
ABSTRACT
Gene duplication events exert key functions on gene innovations during the evolution of the eukaryotic genomes. A large portion of the total gene content in plants arose from tandem duplications events, which often result in paralog genes with high sequence identity. Ubiquitin ligases or E3 enzymes are components of the ubiquitin proteasome system that function during the transfer of the ubiquitin molecule to the substrate. In plants, several E3s have expanded in their genomes as multigene families. To gain insight into the consequences of gene duplications on the expansion and diversification of E3s, we examined the evolutionary basis of a cluster of six genes, duplC-ATLs, which arose from segmental and tandem duplication events in Brassicaceae. The assessment of the expression suggested two patterns that are supported by lineage. While retention of expression domains was observed, an apparent absence or reduction of expression was also inferred. We found that two duplC-ATL genes underwent pseudogenization and that, in one case, gene expression is probably regained. Our findings provide insights into the evolution of gene families in plants, defining key events on the expansion of the Arabidopsis Tóxicos en Levadura family of E3 ligases.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution