Merck
  • Targeting colorectal cancer cells with single-walled carbon nanotubes conjugated to anticancer agent SN-38 and EGFR antibody.
Targeting colorectal cancer cells with single-walled carbon nanotubes conjugated to anticancer agent SN-38 and EGFR antibody.
Biomaterials (2013-08-14)
Pei-Chi Lee, Yu-Chi Chiou, Jau-Min Wong, Cheng-Liang Peng, Ming-Jium Shieh
ABSTRACT
In this study, single-walled carbon nanotubes (SWNTs) conjugated with antibody C225 were used to achieve targeted therapy against EGFR over-expressed colorectal cancer cells. In addition, the control release of the chemotherapeutic drug, 7-Ethyl-10-hydroxy-camptothecin (SN38), was studied. We used three different colorectal cancer cell lines, HCT116, HT29, and SW620, listed in the order of decreasing expression levels of EGFR. Our results showed that SWNT could use C225 to specifically bind to EGFR-expressed cells. The cellular uptakes of SWNT of EGFR over-expressed cells (HCT116 and HT29) were much higher than that of the negative control (SW620). We, next, demonstrated that receptor-mediated endocytosis was the primary cell entry route for SWNT. As a consequence, abundant amount of SN38 was released and EGFR over-expressed cells were killed. The drug control release process was studied by utilizing human carboxylesterase enzyme (hCE) that would break the bond linking SN38 and SWNT-carrier in cytoplasm. The intracellular SN38 release observed by confocal microscopy showed that SN38 actually dissociated from the SWNT-carrier first. SN38's entry to nucleus was then followed while the SWNT-carrier still remained in the cytoplasm. Overall, all these data suggested that SWNT could be a good carrier for targeting controlled release therapy.
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg