Merck
  • In memory of Professor Leonor Michaelis in Nagoya: great contributions to biochemistry in Japan in the first half of the 20th century.
In memory of Professor Leonor Michaelis in Nagoya: great contributions to biochemistry in Japan in the first half of the 20th century.
FEBS letters (2013-05-01)
Toshiharu Toshi Nagatsu
ABSTRACT
Leonor Michaelis spent the years of 1922-1926 as Professor of Biochemistry of the Aichi Medical College (now Graduate School of Medicine, Nagoya University) in Nagoya, Japan. Michaelis succeeded in gathering many bright young biochemists from all over Japan into his laboratory, and made tremendous contributions to the promotion of biochemistry in Japan. Michaelis was invited to many places in Japan to present lectures over those years. Kunio Yagi, who was Professor of Biochemistry at Nagoya University in the second half of the 20th century, succeeded in crystallizing the "Michaelis" enzyme-substrate complex. Historically, Michelis has had an enormous impact on biochemistry in Japan.
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid