Merck
  • BioShuttle mobility in living cells studied with high-resolution FCS & CLSM methodologies.
BioShuttle mobility in living cells studied with high-resolution FCS & CLSM methodologies.
International journal of medical sciences (2012-07-20)
Klaus Braun, Marcel Beining, Manfred Wiessler, Twan Lammers, Rüdiger Pipkorn, Ute Hennrich, Kiyoshi Nokihara, Wolfhard Semmler, Jürgen Debus, Waldemar Waldeck
ABSTRACT
With the increase in molecular diagnostics and patient-specific therapeutic approaches, the delivery and targeting of imaging molecules and pharmacologically active agents gain increasing importance. The ideal delivery system does not exist yet. The realization of two features is indispensable: first, a locally high concentration of target-specific diagnostic and therapeutic molecules; second, the broad development of effective and safe carrier systems. Here we characterize the transport properties of the peptide-based BioShuttle transporter using FFM and CLSM methods. The modular design of BioShuttle-based formulations results in a multi-faceted field of applications, also as a theranostic tool.
Sigma-Aldrich
Atto 488-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488 amine, BioReagent, ≥90% (HPLC), suitable for fluorescence
Sigma-Aldrich
Atto 488 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488 maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)