Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Discovery of potent and orally bioavailable N,N'-diarylurea antagonists for the CXCR2 chemokine receptor.
Discovery of potent and orally bioavailable N,N'-diarylurea antagonists for the CXCR2 chemokine receptor.
Bioorganic & medicinal chemistry letters (2004-09-11)
Qi Jin, Hong Nie, Brent W McCleland, Katherine L Widdowson, Michael R Palovich, John D Elliott, Richard M Goodman, Miriam Burman, Henry M Sarau, Keith W Ward, Melanie Nord, Bonnie M Orr, Peter D Gorycki, Jakob Busch-Petersen
ABSTRACT
A series of 3-substituted N,N'-diarylureas was prepared and the structure-activity relationship relative to CXCR2 receptor affinity as well as their pharmacokinetic properties were examined. In vitro microsomal metabolism studies indicated that the lower clearance rates of the 3-sulfonamido-substituted compounds were most likely due to the suppression of glucuronidation.