Merck
  • In vitro activity of ceftolozane-tazobactam against Pseudomonas aeruginosa isolates obtained from patients in Canadian hospitals in the CANWARD study, 2007 to 2012.
In vitro activity of ceftolozane-tazobactam against Pseudomonas aeruginosa isolates obtained from patients in Canadian hospitals in the CANWARD study, 2007 to 2012.
Antimicrobial agents and chemotherapy (2013-08-14)
A Walkty, J A Karlowsky, H Adam, M Baxter, P Lagacé-Wiens, D J Hoban, G G Zhanel
ABSTRACT
The in vitro activity of ceftolozane in combination with tazobactam (fixed concentration of 4 μg/ml) was evaluated against 2,435 Pseudomonas aeruginosa clinical isolates obtained from across Canada using Clinical and Laboratory Standards Institute broth microdilution methods. The MIC50 and MIC90 values for ceftolozane-tazobactam were 0.5 μg/ml and 1 μg/ml, respectively (a 32-fold-lower MIC90 than that for ceftazidime). Eighty-nine percent (141/158) of multidrug-resistant isolates were inhibited by ≤8 μg/ml of ceftolozane-tazobactam.
Product Description
USP
Ceftazidime pentahydrate, United States Pharmacopeia (USP) Reference Standard
Ceftazidime, European Pharmacopoeia (EP) Reference Standard
Ceftazidime for peak identification, European Pharmacopoeia (EP) Reference Standard