Merck
  • Hypertensive cardiac remodeling effects of lignan extracts from Eucommia ulmoides Oliv. bark--a famous traditional Chinese medicine.
Hypertensive cardiac remodeling effects of lignan extracts from Eucommia ulmoides Oliv. bark--a famous traditional Chinese medicine.
The American journal of Chinese medicine (2013-07-31)
Zhen-Yu Li, Juan Gu, Jin Yan, Jun-Jie Wang, Wei-Hua Huang, Zhi-Rong Tan, Gan Zhou, Yao Chen, Hong-Hao Zhou, Dong-Sheng Ouyang
ABSTRACT
The lignan extracts from the tree bark of Eucommia ulmoides Oliv., a famous traditional Chinese medicine, have been demonstrated to have inhibitory effects on aldose reductase activity in spontaneously hypertensive rat myocardium. This study was aimed to investigate the hypertensive cardiac remodeling effects of the lignan extracts together with epalrestat. Ten-week-old male spontaneously hypertensive rats were randomly divided into three groups (n = 12, each) and administered 100 mg/kg/d of captopril (angiotensin converting enzyme inhibitor), 100 mg/kg/d of epalrestat (aldose reductase inhibitor) or 300 mg/kg/d of lignan extracts by gavage for 16 weeks. Sex-, age-, and number-matched normotensive Wistar Kyoto rats with spontaneously hypertensive rats were treated with distilled water (vehicle) as controls. Systolic blood pressures were measured periodically. Echocardiography examination was taken when rats were 24 weeks old. We found that both captopril and lignan extracts lowered blood pressure, and inhibited aldose reductase activity similarly to epalrestat. Echocardiography examination and histomorphometry indices were improved in all treated groups (p < 0.05). Therefore, lignan extracts could prevent hypertensive cardiac remodeling, which is likely related to aldose reductase inhibition.
Sigma-Aldrich
Epalrestat, ≥98% (HPLC)
Pricing and availability is not currently available.