Merck
  • Cefpodoxime proxetil-related hemolysis and acute interstitial nephritis.
Cefpodoxime proxetil-related hemolysis and acute interstitial nephritis.
Current drug safety (2013-07-13)
Hakki Yilmaz, Mukadder Ayse Bilgiç, Nuket Bavbek, Ali Akçay
ABSTRACT
We report a case of acute interstitial nephritis (AIN) and immune hemolytic anemia (IHA) associated with cefpodoxime therapy. A patient with a recent history of cefpodoxime proxetil treatment presented with elevated serum creatinine, oliguria, nausea, vomiting, and dyspnea. Evidence of renal failure, abnormal urinalysis, and renal biopsy with inflammatory infiltrate in the interstitium confirmed a diagnosis of AIN. The patient subsequently developed IHA, which was confirmed by peripheral blood smear results and positive Coombs' test. The patient recovered after dialysis therapy and 2 days of intravenous methylprednisolone (500mg/day) followed by oral prednisolone (60 mg/day), which was rapidly tapered and stopped within 3 weeks. To our knowledge, cefpodoxime-induced AIN and IHA are unprecedented. Physicians should be aware that drug-induced AIN and hemolysis can be associated with cefpodoxime proxetil.
Product Description
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Cefpodoxime proxetil, United States Pharmacopeia (USP) Reference Standard
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard