Merck
[Research on quality standards of herbs of Peganum harmala].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-12-29)
Fang-fang Wen, Li-ming Zheng, Xiao-jing Li, Yan Li, Lei Zhang, Xue-mei Cheng, Chang-hong Wang, Zheng-taou Wang
ABSTRACT
To establish the quality standards of the herbs of Peganum harmala. According to the Chinese Pharmacopoeia (2010 version, volume 1) and its appendix method, the water, total ash, acid insoluble ash, water-soluble extractives, and heavy metal were analyzed for herbs of P. harmala. TLC method was used to separate harmaline, harmine and vasicine from the herb samples by mixture of ethyl acetate-methanol-ammonia (10: 1.5: 0.5) as a developing solvent on high performance silica gel precoated plate (HSGF254) and to identify them inspected under UV 366 nm, visualized by spraying with both Dragendorff reagent, and by bioautographic assay. In the HPLC method, vasicine was separated on a C18 (4.6 mm x 250 mm, 5 microm) column with metnanol-0.1% trifluoroacetic acid (15:85) as the mobile phase and detected at at 280 nm. In the TLC procedures, 254 nm fluorescent and bioautographic assay for the detection of acetylcholinesterase inhibitor can be used for the qualitative identification of the active ingredients. For the HPLC quantitation method, the calibration curve of vasicine displayed ideal linearity over the range of 0.7923-792.3 mg x L(-1) with the regression equation of Y = 18,227X - 24.879 (r = 0.9999). The average recovery of vasicine was 101.6% with a RSD of 1.9%. The RSD values of intra-day and inter-day precision were less than 2%. The content of vasicine in 10 batches of herbs of P. harmala fluctuates between 0.23% and 1.47%. The results indicated that the limit of vasicine was not lower than 0.6%, and the water, total ash, acid insoluble ash, and water-soluble extractives were not more than 10.0%, 20.0%, 1.7%, and 30.0%, respectively. The heavy metal of plumbum, cadmium, arsenic, mercury, and copper were not more than 5, 3, 2, 2, and 20 mg x kg(-1), respectively. The qualitative and quantitative method established was suitable for the quality evaluation and assessment of herbs of P. harmala.