- Single dose therapy of vaginal candidiasis: a comparative trial of fenticonazole vaginal ovules versus clotrimazole vaginal tablets.
Single dose therapy of vaginal candidiasis: a comparative trial of fenticonazole vaginal ovules versus clotrimazole vaginal tablets.
An open, randomized comparative clinical trial was performed in 153 patients suffering from symptomatic vaginal candidiasis confirmed by mycological tests. Patients were allocated at random into two groups: the first group (consisting of 75 subjects) was treated with a single vaginal ovule of fenticonazole (600 mg) and the second group (consisting of 78 subjects) was treated with a single vaginal tablet of clotrimazole (500 mg). Therapeutic efficacy was assessed by microbiological and clinical criteria 7 days and 1 month (when possible) after the single dose treatment. At the first follow-up visit, complete disappearance of the signs and symptoms or a highly significant reduction of their intensity was observed in both treatment groups. No significant difference was evident between the two drugs. At 7 days, the mycological tests gave negative results in 92% of the patients in the fenticonazole group and in 88.5% of the patients in the clotrimazole group. The difference between the two treatment groups was again not statistically significant. The second follow-up visit was performed in 55 (73.3%) patients of the fenticonazole group and in 52 (66.7%) patients of the clotrimazole group. The results indicate that 83.6% of patients in the fenticonazole group and 69.2% of patients in the clotrimazole group were still disease free at the time of this visit. Both drugs were well tolerated. Mild, local and short lasting side-effects were reported in only 5 cases of the group treated with fenticonazole.(ABSTRACT TRUNCATED AT 250 WORDS)