Merck
  • Investigation of the lipophilic behaviour of some thiazolidinediones. Relationships with PPAR-gamma activity.
Investigation of the lipophilic behaviour of some thiazolidinediones. Relationships with PPAR-gamma activity.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2007-07-31)
Costas Giaginis, Stamatios Theocharis, Anna Tsantili-Kakoulidou
ABSTRACT
Various lipophilicity aspects of five well-known PPAR-gamma ligands, belonging to the thiazolidinedione (TZD) class, ciglitazone (CSZ), troglitazone (TGZ), netoglitazone (NGZ) and the ampholytic pioglitazone (PGZ) and rosiglitazone (RGZ), have been explored. The compounds were found to be highly lipophilic as assessed by direct octanol-water partitioning experiments and further confirmed by reversed phase HPLC measurements under different conditions. Immobilised artificial membrane (IAM) chromatographic indices were also determined as an alternative expression of lipophilicity. They were found to show less diversity forming two clusters. Experimental logD/logP values were compared to those predicted by three widely used calculation systems. For the two ampholytic TZDs, the lipophilicity and retention/pH profiles were established over a broad pH range and compared to the corresponding calculated profiles. Lipophilicity indices derived under the different conditions were further compared to biological activity, concerning in vitro transactivation (pEC(50)) and binding affinity (pK(i)) data, taken from literature. The most active TZD (RGZ) in both transactivation and binding assay proved to be the less lipophilic analogue. An equation relating pEC(50) data to experimental logD(7.4) or reversed-phase logk(w) values could be established, while pK(i) data did not lead to satisfactory correlation.
Sigma-Aldrich
MCC-555, ≥98% (HPLC)