Merck
  • In vivo analysis of metabolic dynamics in Saccharomyces cerevisiae : I. Experimental observations.
In vivo analysis of metabolic dynamics in Saccharomyces cerevisiae : I. Experimental observations.
Biotechnology and bioengineering (1997-07-20)
U Theobald, W Mailinger, M Baltes, M Rizzi, M Reuss
ABSTRACT
The goal of this work was to obtain rapid sampling technique to measure transient metabolites in vivo. First, a pulse of glucose was added to a culture of the yeast Saccharomyces cerevisiae growing aerobically under glucose limitation. Next, samples were removed at 2 to 5 s intervals and quenched using methods that depend on the metabolite measured. Extracellular glucose, excreted products, as well as glycolytic intermediates (G6P, F6P, FBP, GAP, 3-PG, PEP, Pyr) and cometabolites (ATP, ADP, AMP, NAD(+), NADH) were measured using enzymatic or HPLC methods. Significant differences between the adenine nucleotide concentrations in the cytoplasm and mitochondria indicated the importance of compartmentation for the regulation of the glycolysis. Changes in the intra- and extracellular levels of metabolites confirmed that glycolysis is regulated on a time scale of seconds. (c) 1997 John Wiley & Sons, Inc. Biotechnol Bioeng 55: 305-316, 1997.
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt, ≥98% (enzymatic)
Sigma-Aldrich
Phospho(enol)pyruvic acid monosodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Phospho(enol)pyruvic acid cyclohexylammonium salt, ≥97% (enzymatic)