Merck
  • A trial of treatment for acute otorrhea in children with tympanostomy tubes.
A trial of treatment for acute otorrhea in children with tympanostomy tubes.
The New England journal of medicine (2014-02-21)
Thijs M A van Dongen, Geert J M G van der Heijden, Roderick P Venekamp, Maroeska M Rovers, Anne G M Schilder
ABSTRACT
Recent guidance for the management of acute otorrhea in children with tympanostomy tubes is based on limited evidence from trials comparing oral antibiotic agents with topical antibiotics. In this open-label, pragmatic trial, we randomly assigned 230 children, 1 to 10 years of age, who had acute tympanostomy-tube otorrhea to receive hydrocortisone-bacitracin-colistin eardrops (76 children) or oral amoxicillin-clavulanate suspension (77) or to undergo initial observation (77). The primary outcome was the presence of otorrhea, as assessed otoscopically, 2 weeks after study-group assignment. Secondary outcomes were the duration of the initial otorrhea episode, the total number of days of otorrhea and the number of otorrhea recurrences during 6 months of follow-up, quality of life, complications, and treatment-related adverse events. Antibiotic-glucocorticoid eardrops were superior to oral antibiotics and initial observation for all outcomes. At 2 weeks, 5% of children treated with antibiotic-glucocorticoid eardrops had otorrhea, as compared with 44% of those treated with oral antibiotics (risk difference, -39 percentage points; 95% confidence interval [CI], -51 to -26) and 55% of those treated with initial observation (risk difference, -49 percentage points; 95% CI, -62 to -37). The median duration of the initial episode of otorrhea was 4 days for children treated with antibiotic-glucocorticoid eardrops versus 5 days for those treated with oral antibiotics (P<0.001) and 12 days for those who were assigned to initial observation (P<0.001). Treatment-related adverse events were mild, and no complications of otitis media, including local cellulitis, perichondritis, mastoiditis, and intracranial complications, were reported at 2 weeks. Antibiotic-glucocorticoid eardrops were more effective than oral antibiotics and initial observation in children with tympanostomy tubes who had uncomplicated acute otorrhea. (Funded by the Netherlands Organization for Health Research and Development; Netherlands Trial Register number, NTR1481.).
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bacitracin zinc salt, from Bacillus licheniformis, ~70,000 U/g
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture