Merck
  • Accuracy and reproducibility of dental replica models reconstructed by different rapid prototyping techniques.
Accuracy and reproducibility of dental replica models reconstructed by different rapid prototyping techniques.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics (2014-01-01)
Aletta Hazeveld, James J R Huddleston Slater, Yijin Ren
ABSTRACT
Rapid prototyping is a fast-developing technique that might play a significant role in the eventual replacement of plaster dental models. The aim of this study was to investigate the accuracy and reproducibility of physical dental models reconstructed from digital data by several rapid prototyping techniques. Twelve mandibular and maxillary conventional plaster models from randomly chosen subjects were selected and served as the gold standard. The plaster models were scanned to form high-resolution 3-dimensional surface models in .stl files. These files were converted into physical models using 3 rapid prototyping techniques: digital light processing, jetted photopolymer, and 3-dimensional printing. Linear measurements on the plaster models were compared with linear measurements on the rapid prototyping models. One observer measured the height and width of the clinical crowns of all teeth (first molar to first molar) on all models (plaster and replicas) using a digital caliper. All models were measured 5 times with a 2-week interval between measurements. The intraobserver agreement was high (intraclass correlation coefficient >0.94). The mean systematic differences for the measurements of the height of the clinical crowns were -0.02 mm for the jetted photopolymer models, 0.04 mm for the digital light processing models, and 0.25 mm for the 3-dimensional printing models. For the width of the teeth, the mean systematic differences were -0.08 mm for the jetted photopolymer models, -0.05 mm for the digital light processing models, and -0.05 mm for the 3-dimensional printing models. Dental models reconstructed by the tested rapid prototyping techniques are considered clinically acceptable in terms of accuracy and reproducibility and might be appropriate for selected applications in orthodontics.
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%