Merck
  • Postural orthostatic tachycardia syndrome (POTS) and vitamin B12 deficiency in adolescents.
Postural orthostatic tachycardia syndrome (POTS) and vitamin B12 deficiency in adolescents.
Pediatrics (2013-12-25)
Taliha Öner, Baris Guven, Vedide Tavli, Timur Mese, Murat Muhtar Yilmazer, Savas Demirpence
ABSTRACT
Vitamin B12 is involved in the production of adrenaline from noradrenaline. It is the cofactor involved in catecholamine degradation and plays a role in myelin synthesis. The current study aimed to investigate the association between vitamin B12 levels and postural orthostatic tachycardia syndrome (POTS) during adolescence when accelerated myelin synthesis increases the vitamin B12 need. One hundred twenty-five patients (mean age 11.1 ± 2.3 years; 60% female) reporting short-term loss of consciousness and diagnosed with vasovagal syncope based on anamnesis with a normal distribution and 50 control subjects (mean age 10.94 ± 2.5 years, 62% female) were included in this study. Serum vitamin B12, folic acid, and ferritin levels were measured prospectively in addition to other tests. We defined vitamin B12 deficiency as a serum level <300 pg/mL.(1-4) RESULTS: Vitamin B12 levels were significantly lower in the patient group compared with the control group (47.2% vs 18%, P < .001). In the patient group, children with the POTS pattern had significantly lower vitamin B12 levels compared with children without the POTS response (P = .03). Vitamin B12 deficiency in patients with POTS may lead to sympathetic nervous system baroreceptor dysfunction.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material