Merck
  • Method for detecting the reactivity of chemicals towards peptides as an alternative test method for assessing skin sensitization potential.
Method for detecting the reactivity of chemicals towards peptides as an alternative test method for assessing skin sensitization potential.
Toxicology letters (2013-12-24)
Sun-A Cho, Yun Hyeok Jeong, Ji Hoon Kim, Seoyoung Kim, Jun-Cheol Cho, Yong Heo, Young Heo, Kyung-Do Suh, Kyeho Shin, Susun An
ABSTRACT
Cosmetics are normally composed of various ingredients. Some cosmetic ingredients can act as chemical haptens reacting toward proteins or peptides of human skin and they can provoke an immunologic reaction, called as skin sensitization. This haptenation process is very important step of inducing skin sensitization and evaluating the sensitizing potentials of cosmetic ingredients is very important for consumer safety. Therefore, animal alternative methods focusing on monitoring haptenation potential are undergoing vigorous research. To examine the further usefulness of spectrophotometric methods to monitor reactivity of chemicals toward peptides for cosmetic ingredients. Forty chemicals (25 sensitizers and 15 non-sensitizers) were reacted with 2 synthetic peptides, e.g., the cysteine peptides (Ac-RFAACAA-COOH) with free thiol group and the lysine peptides (Ac-RFAAKAA-COOH) with free amine group. Unreacted peptides can be detected after incubating with 5,5'-dithiobis-2-nitrobenzoic acid or fluorescamine™ as detection reagents for free thiol and amine group, respectively. Chemicals were categorized as sensitizers when they induced more than 10% depletion of cysteine peptides or more than 30% depletion of lysine peptides. The sensitivity, specificity, and accuracy were 80.0%, 86.7% and 82.5%, respectively. These results demonstrate that spectrophotometric methods can be an easy, fast, and high-throughput screening tools predicting the skin sensitization potential of chemical including cosmetic ingredient.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine, Vetec, reagent grade, 97%
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%