Merck
Targeted polysaccharide nanoparticle for adamplatin prodrug delivery.
Journal of medicinal chemistry (2013-11-21)
Yang Yang, Ying-Ming Zhang, Yong Chen, Jia-Tong Chen, Yu Liu
ABSTRACT
A series of conjugated hyaluronic acid particles (HAP), composed of a hydrophobic anticancer drug core and hydrophilic cyclodextrin/hyaluronic acid shell, were prepared through self-assembling and characterized by (1)H NMR titration, electron microscopy, zeta potential, and dynamic light-scattering experiments. The nanometer-sized HAP thus prepared was biocompatible and biodegradable and was well-recognized by the hyaluronic acid receptors overexpressed on the surface of cancer cells, which enabled us to exploit HAP as an efficient targeted delivery system for anticancer drugs. Indeed, HAP exhibited anticancer activities comparable to the commercial anticancer drug cisplatin but with lower side effects both in vitro and in vivo.
Sigma-Aldrich
Adamantane, ≥99%
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000