Merck
Clinical practice: Adult attention deficit-hyperactivity disorder.
The New England journal of medicine (2013-11-15)
Nora D Volkow, James M Swanson
ABSTRACT
Short-term trials involving adults with ADHD have shown significant improvements in symptoms with stimulants and atomoxetine; however, data on long-term benefits and risks of these medications, particularly among older persons, have been insufficient.
Sigma-Aldrich
(R)-Tomoxetine hydrochloride, solid
Pricing and availability is not currently available.