Merck
TSPAN33 is a novel marker of activated and malignant B cells.
Clinical immunology (Orlando, Fla.) (2013-11-12)
Van Phi Luu, Peter Hevezi, Felipe Vences-Catalan, José Luis Maravillas-Montero, Clayton Alexander White, Paolo Casali, Luis Llorente, Juan Jakez-Ocampo, Guadalupe Lima, Natalia Vilches-Cisneros, Juan Pablo Flores-Gutiérrez, Leopoldo Santos-Argumedo, Albert Zlotnik
ABSTRACT
We have identified Tspan33 as a gene encoding a transmembrane protein exhibiting a restricted expression pattern including expression in activated B cells. TSPAN33 is a member of the tetraspanin family. TSPAN33 is not expressed in resting B cells, but is strongly induced in primary human B cells following activation. Human 2E2 cells, a Burkitt's lymphoma-derived B cell model of activation and differentiation, also upregulate TSPAN33 upon activation. TSPAN33 is expressed in several lymphomas including Hodgkin's and Diffuse large B cell lymphoma. TSPAN33 is also expressed in some autoimmune diseases where B cells participate in the pathology, including rheumatoid arthritis patients, systemic lupus erythematosus (SLE), and in spleen B cells from MRL/Fas(lpr/lpr) mice (a mouse model of SLE). We conclude that TSPAN33 may be used as a diagnostic biomarker or as a target for therapeutic antibodies for treatment of certain B cell lymphomas or autoimmune diseases.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
4α-Phorbol 12-myristate 13-acetate, solid, ≥95% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder