Merck
  • Adverse reaction to metal debris after ReCap-M2A-Magnum large-diameter-head metal-on-metal total hip arthroplasty.
Adverse reaction to metal debris after ReCap-M2A-Magnum large-diameter-head metal-on-metal total hip arthroplasty.
Acta orthopaedica (2013-11-01)
Jari Mokka, Mika Junnila, Matti Seppänen, Petri Virolainen, Tuukka Pölönen, Tero Vahlberg, Kimmo Mattila, Esa K J Tuominen, Juho Rantakokko, Ville Aärimaa, Juha Kukkonen, Keijo T Mäkelä
ABSTRACT
The clinical findings of adverse reaction to metal debris (ARMD) following large-diameter-head metal-on-metal total hip arthroplasty (LDH MoM THA) may include periarticular fluid collections, soft tissue masses, and gluteal muscle necrosis. The ReCap-M2a-Magnum LDH MoM THA was the most commonly used hip device at our institution from 2005 to 2012. We assessed the prevalence of and risk factors for ARMD with this device. 74 patients (80 hips) had a ReCap-M2a-Magnum LDH MoM THA during the period August 2005 to December 2006. These patients were studied with hip MRI, serum chromium and cobalt ion measurements, the Oxford hip score questionnaire, and by clinical examination. The prevalence of ARMD was recorded and risk factors for ARMD were assessed using logistic regression models. The mean follow-up time was 6.0 (5.5-6.7) years. A revision operation due to ARMD was needed by 3 of 74 patients (3 of 80 hips). 8 additional patients (8 hips) had definite ARMD, but revision was not performed. 29 patients (32 hips) were considered to have a probable or possible ARMD. Altogether, 43 of 80 hips had a definite, probable, or possible ARMD and 34 patients (37 hips) were considered not to have ARMD. In 46 of 78 hips, MRI revealed a soft tissue mass or a collection of fluid (of any size). The symptoms clicking in the hip, local hip swelling, and a feeling of subluxation were associated with ARMD. ARMD is common after ReCap-M2a-Magnum total hip arthroplasty, and we discourage the use of this device. Asymptomatic patients with a small fluid collection on MRI may not need instant revision surgery but must be followed up closely.
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis