Merck
  • Sudden death from toluene intoxication: a case report and review of literature.
Sudden death from toluene intoxication: a case report and review of literature.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-10-30)
Weerapong Prayulsatien
ABSTRACT
Toluene is an aromatic hydrocarbon (C7H8) found in gasoline, acrylic paints, varnishes, lacquers, paint thinners, adhesives, and shoe polish. Toluene toxicity can occur from unintentional or deliberate inhalation of fumes, ingestion, or transdermal absorption. Unintentional exposure to high concentrations of toluene results in severe toluene intoxication. This is a report of toluene poisoning in a middle-age man found dead at home after varnish application by spray. There was no obvious external and internal cause of death. The blood test revealed the presence of toluene. The cause of death was diagnosed as cardiopulmonary failure caused by toluene, which is a rare case report in Thailand.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)