Merck
  • Reduced-intensity conditioning and HLA-matched haemopoietic stem-cell transplantation in patients with chronic granulomatous disease: a prospective multicentre study.
Reduced-intensity conditioning and HLA-matched haemopoietic stem-cell transplantation in patients with chronic granulomatous disease: a prospective multicentre study.
Lancet (London, England) (2013-10-29)
Tayfun Güngör, Pierre Teira, Mary Slatter, Georg Stussi, Polina Stepensky, Despina Moshous, Clementien Vermont, Imran Ahmad, Peter J Shaw, José Marcos Telles da Cunha, Paul G Schlegel, Rachel Hough, Anders Fasth, Karim Kentouche, Bernd Gruhn, Juliana F Fernandes, Silvy Lachance, Robbert Bredius, Igor B Resnick, Bernd H Belohradsky, Andrew Gennery, Alain Fischer, H Bobby Gaspar, Urs Schanz, Reinhard Seger, Katharina Rentsch, Paul Veys, Elie Haddad, Michael H Albert, Moustapha Hassan
ABSTRACT
In chronic granulomatous disease allogeneic haemopoietic stem-cell transplantation (HSCT) in adolescents and young adults and patients with high-risk disease is complicated by graft-failure, graft-versus-host disease (GVHD), and transplant-related mortality. We examined the effect of a reduced-intensity conditioning regimen designed to enhance myeloid engraftment and reduce organ toxicity in these patients. This prospective study was done at 16 centres in ten countries worldwide. Patients aged 0-40 years with chronic granulomatous disease were assessed and enrolled at the discretion of individual centres. Reduced-intensity conditioning consisted of high-dose fludarabine (30 mg/m(2) [infants <9 kg 1·2 mg/kg]; one dose per day on days -8 to -3), serotherapy (anti-thymocyte globulin [10 mg/kg, one dose per day on days -4 to -1; or thymoglobuline 2·5 mg/kg, one dose per day on days -5 to -3]; or low-dose alemtuzumab [<1 mg/kg on days -8 to -6]), and low-dose (50-72% of myeloablative dose) or targeted busulfan administration (recommended cumulative area under the curve: 45-65 mg/L × h). Busulfan was administered mainly intravenously and exceptionally orally from days -5 to -3. Intravenous busulfan was dosed according to weight-based recommendations and was administered in most centres (ten) twice daily over 4 h. Unmanipulated bone marrow or peripheral blood stem cells from HLA-matched related-donors or HLA-9/10 or HLA-10/10 matched unrelated-donors were infused. The primary endpoints were overall survival and event-free survival (EFS), probabilities of overall survival and EFS at 2 years, incidence of acute and chronic GVHD, achievement of at least 90% myeloid donor chimerism, and incidence of graft failure after at least 6 months of follow-up. 56 patients (median age 12·7 years; IQR 6·8-17·3) with chronic granulomatous disease were enrolled from June 15, 2003, to Dec 15, 2012. 42 patients (75%) had high-risk features (ie, intractable infections and autoinflammation), 25 (45%) were adolescents and young adults (age 14-39 years). 21 HLA-matched related-donor and 35 HLA-matched unrelated-donor transplants were done. Median time to engraftment was 19 days (IQR 16-22) for neutrophils and 21 days (IQR 16-25) for platelets. At median follow-up of 21 months (IQR 13-35) overall survival was 93% (52 of 56) and EFS was 89% (50 of 56). The 2-year probability of overall survival was 96% (95% CI 86·46-99·09) and of EFS was 91% (79·78-96·17). Graft-failure occurred in 5% (three of 56) of patients. The cumulative incidence of acute GVHD of grade III-IV was 4% (two of 56) and of chronic graft-versus-host disease was 7% (four of 56). Stable (≥90%) myeloid donor chimerism was documented in 52 (93%) surviving patients. This reduced-intensity conditioning regimen is safe and efficacious in high-risk patients with chronic granulomatous disease. None.
Sigma-Aldrich
Adenine 9-β-D-arabinofuranoside, ≥99%
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor
Busulfan, European Pharmacopoeia (EP) Reference Standard
Supelco
Busulfan, analytical standard, for drug analysis