Merck
  • N-hexane alters the maturation of oocytes and induces apoptosis in mice.
N-hexane alters the maturation of oocytes and induces apoptosis in mice.
Biomedical and environmental sciences : BES (2013-10-09)
Jin Liu, Lei Huang, Yan Sun, Yu Chen Li, Jian Lin Zhu, Wen Xiang Wang, Wen Chang Zhang
ABSTRACT
This study was aimed to determine the effects of n-hexane on the maturation of mouse oocytes. Cell culture was used to observe the maturation of mouse oocytes and CLSM was employed to determine their apoptosis. Germinal vesicle breakdown (GVBD) and extrusion of the first polar body in mouse oocytes were significantly inhibited by n-hexane. After fertilization, the number of eggs in the mouse was significantly reduced by n-hexane. Mitochondrial membrane potentials (ΔΨm) were altered in mouse oocytes that were leading to apoptosis of the oocytes. N-hexane might have affected the maturation of oocytes, causing alteration of ΔΨm and leading to apoptosis which maybe one of the most important mechanisms.
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%