Merck
  • A phase 2 trial of azacitidine and gemtuzumab ozogamicin therapy in older patients with acute myeloid leukemia.
A phase 2 trial of azacitidine and gemtuzumab ozogamicin therapy in older patients with acute myeloid leukemia.
Blood (2013-10-05)
Sucha Nand, Megan Othus, John E Godwin, Cheryl L Willman, Thomas H Norwood, Dianna S Howard, Steven E Coutre, Harry P Erba, Frederick R Appelbaum
ABSTRACT
This trial tested the safety and efficacy of a regimen consisting of hydroxyurea followed by azacitidine, 75 mg/m(2) for 7 days, and gemtuzumab ozogamicin, 3 mg/m(2) on day 8, in older patients with newly diagnosed acute myeloid leukemia. Those achieving a complete remission received 1 consolidation treatment followed by 4 cycles of azacitidine. The patients were stratified into good-risk (age 60-69 years or performance status 0-1) and poor-risk (age ≥70 years and performance status 2 or 3) groups. Specific efficacy and safety goals were defined as being supportive of further study of the regimen. Eighty-three patients were registered in the good-risk cohort and 59 in poor-risk cohort, with median age of 71 and 75 years, respectively. In the good-risk group, 35 patients (44%) achieved a complete remission. Median relapse-free and overall survivals were 8 and 11 months, respectively. Six patients (8%) died within 30 days of registration. In the poor-risk group, 19 (35%) achieved a complete remission. Median relapse-free and overall survivals were 7 and 11 months, respectively. Seven patients (14%) died early. The results of this trial met predefined goals for efficacy and safety for the poor-risk cohort but not the good-risk group. .
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)