Merck
  • A facile label-free electrochemiluminescence biosensor for target protein specific recognition based on the controlled-release delivery system.
A facile label-free electrochemiluminescence biosensor for target protein specific recognition based on the controlled-release delivery system.
Talanta (2013-08-21)
Xiaoyan Yang, Aiguo Wang, Jilong Liu
ABSTRACT
This paper described a novel label-free electrochemiluminescence assay for target protein based on a controlled delivery system. Iron oxide magnetic mesoporous silica nanocontainers were prepared by using a general procedure. The prepared magnetic mesoporous silica nanocontainers were applied to load the guest molecules [Ru(bpy)3](2+). Aptamers were used as gatekeepers on the pore outlets of the nanocontainers. In the presence of target proteins, the specific aptamer-protein interactions were employed as triggers for uncapping the pores and releasing the guest molecules from the nanocontainers. The amount of the guest molecule [Ru(bpy)3](2+) released from the magnetic mesoporous silica nanocontainers was monitored by the electrochemiluminescence assay. The results show that the releasing amount of [Ru(bpy)3](2+) is proportional to the thrombin concentration in the range of 0.6 pM-0.8 nM with a detection limit of 0.5 pM (S/N=3). The present work demonstrates that the fabricated nanocontainer using aptamer as the cap is a highly sensitive and selective key-in lock gating system for the label-free ECL biosensor.
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene