Merck
  • Overexpression of catalase in vascular smooth muscle cells prevents the formation of abdominal aortic aneurysms.
Overexpression of catalase in vascular smooth muscle cells prevents the formation of abdominal aortic aneurysms.
Arteriosclerosis, thrombosis, and vascular biology (2013-08-21)
Ioannis Parastatidis, Daiana Weiss, Giji Joseph, W Robert Taylor
ABSTRACT
Elevated levels of oxidative stress have been reported in abdominal aortic aneurysms (AAA), but which reactive oxygen species promotes the development of AAA remains unclear. Here, we investigate the effect of hydrogen peroxide (H2O2)-degrading enzyme catalase on the formation of AAA. AAA were induced with the application of calcium chloride (CaCl2) on mouse infrarenal aortas. The administration of PEG-catalase, but not saline, attenuated the loss of tunica media and protected against AAA formation (0.91 ± 0.1 versus 0.76 ± 0.09 mm). Similarly, in a transgenic mouse model, catalase overexpression in the vascular smooth muscle cells preserved the thickness of tunica media and inhibited aortic dilatation by 50% (0.85 ± 0.14 versus 0.57 ± 0.08 mm). Further studies showed that injury with CaCl2 decreased catalase expression and activity in the aortic wall. Pharmacological administration or genetic overexpression of catalase restored catalase activity and subsequently decreased matrix metalloproteinase activity. In addition, a profound reduction in inflammatory markers and vascular smooth muscle cell apoptosis was evident in aortas of catalase-overexpressing mice. Interestingly, as opposed to infusion of PEG-catalase, chronic overexpression of catalase in vascular smooth muscle cells did not alter the total aortic H2O2 levels. The data suggest that a reduction in aortic wall catalase activity can predispose to AAA formation. Restoration of catalase activity in the vascular wall enhances aortic vascular smooth muscle cell survival and prevents AAA formation primarily through modulation of matrix metalloproteinase activity.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis