Merck
  • Silencing of TaBTF3 gene impairs tolerance to freezing and drought stresses in wheat.
Silencing of TaBTF3 gene impairs tolerance to freezing and drought stresses in wheat.
Molecular genetics and genomics : MGG (2013-08-15)
Guozhang Kang, Hongzhen Ma, Guoqin Liu, Qiaoxia Han, Chengwei Li, Tiancai Guo
ABSTRACT
Basic transcription factor 3 (BTF3), the β-subunit of the nascent polypeptide-associated complex, is responsible for the transcriptional initiation of RNA polymerase II and is also involved in cell apoptosis, translation initiation regulation, growth, development, and other functions. Here, we report the impact of BTF3 on abiotic tolerance in higher plants. The transcription levels of the TaBTF3 gene, first isolated from wheat seedlings in our lab, were differentially regulated by diverse abiotic stresses and hormone treatments, including PEG-induced stress (20 % polyethylene glycol 6000), cold (4 °C), salt (100 mM NaCl), abscisic acid (100 μM), methyl jasmonate (50 μM), and salicylic acid (50 μM). Southern blot analysis indicated that, in the wheat genome, TaBTF3 is a multi-copy gene. Compared to BSMV-GFP-infected wheat plants (control), under freezing (-8 °C for 48 h) or drought stress (withholding water for 15 days) conditions, TaBTF3-silenced wheat plants showed lower survival rates, free proline content, and relative water content and higher relative electrical conductivity and water loss rate. These results suggest that silencing of the TaBTF3 gene may impair tolerance to freezing and drought stresses in wheat and that it may be involved in the response to abiotic stresses in higher plants.
Product Description
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland