Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition of the tumour necrosis factor-alpha autocrine loop enhances the sensitivity of multiple myeloma cells to anticancer drugs.
Inhibition of the tumour necrosis factor-alpha autocrine loop enhances the sensitivity of multiple myeloma cells to anticancer drugs.
European journal of cancer (Oxford, England : 1990) (2013-08-13)
Masanobu Tsubaki, Makiko Komai, Tatsuki Itoh, Motohiro Imano, Kotaro Sakamoto, Hirotaka Shimaoka, Naoki Ogawa, Kenji Mashimo, Daichiro Fujiwara, Tomoya Takeda, Junji Mukai, Katsuhiko Sakaguchi, Takao Satou, Shozo Nishida
ABSTRACT
Several autocrine soluble factors, including macrophage inflammatory protein-1α and tumour necrosis factor-alpha (TNF-α), promote the survival and growth of multiple myeloma (MM) cells. We hypothesised that inhibition of the TNF-α autocrine loop may enhance the cytotoxic effect of anticancer drugs in MM cell lines. In the present study, a TNF-α-neutralizing antibody suppressed cell proliferation and enhanced the cytotoxic effect of anticancer drugs on MM cells. In addition, combination treatment with the TNF-α-neutralizing antibody and the chemotherapy agent melphalan inhibited nuclear factor κB (NF-κB) p65 nuclear translocation and mammalian target of rapamycin (mTOR) activation and upregulated the expression of Bax and Bim. Treatment of ARH-77 cells with the NF-κB inhibitor dimethyl fumarate or the mTOR inhibitor rapamycin suppressed NF-κB p65 nuclear translocation and enhanced the cytotoxic effect of melphalan. Furthermore, infliximab, a monoclonal antibody against TNF-α, also enhanced the cytotoxic effect of anticancer drugs in ARH-77 cells. These results indicated that TNF-α-neutralizing antibodies or infliximab enhanced the cytotoxic effect of anticancer drugs by suppressing the TNF receptor/mTOR/NF-κB pathways. The inhibition of TNF-α may thus provide a new therapeutic approach to control tumour progression and bone destruction in MM patients.
Sigma-Aldrich
Dimethyl fumarate, 97%
Supelco
Dimethyl fumarate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland