Merck
  • Prophylactic compared with therapeutic ibuprofen analgesia in first-trimester medical abortion: a randomized controlled trial.
Prophylactic compared with therapeutic ibuprofen analgesia in first-trimester medical abortion: a randomized controlled trial.
Obstetrics and gynecology (2013-08-08)
Elizabeth G Raymond, Mark A Weaver, Karmen S Louie, Gillian Dean, Lauren Porsch, E Steve Lichtenberg, Rose Ali, Michelle Arnesen
ABSTRACT
To compare the effectiveness of two oral analgesic regimens in first-trimester medical abortion. We randomly assigned 250 participants undergoing first-trimester abortion with mifepristone and misoprostol at three clinics to two ibuprofen regimens: therapeutic (800 mg every 4-6 hours as needed for pain) or prophylactic (800 mg starting 1 hour before the misoprostol dose, then every 4-6 hours for 48 hours regardless of pain, then as needed). We asked each participant to record her maximum pain on a scale of 0-10 daily thereafter. Of participants assigned to the prophylactic and therapeutic regimens, 111 of 123 (90%) and 117 of 127 (92%), respectively, provided follow-up data. More than 80% of the participants in each group complied with their assigned treatment. Participants in the prophylactic group used substantially more ibuprofen than those in the therapeutic group (median of nine and four tablets, respectively). The mean maximum pain score was 7.1 in the prophylactic group and 7.3 in the therapeutic group (standard deviations 2.5 and 2.2, respectively); the difference was not statistically significant (P=.87, adjusted for site). Duration of pain, verbal pain ratings reported at follow-up, and use of other analgesics did not differ significantly by group (all P>.05). No significant benefit of the prophylactic regimen was apparent in any population subgroup. Abortion failure and ibuprofen side effects in the two groups were similar. We found no evidence that prophylactic administration of ibuprofen reduces pain severity or duration in first-trimester medical abortion. The average pain severity experienced by participants using both regimens was high. ClinicalTrials.gov, www.clinicaltrials.gov, NCT01457521. I.
Product Description
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Misoprostol, ≥99% (HPLC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Misoprostol for system suitability, European Pharmacopoeia (EP) Reference Standard
Misoprostol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%