Merck
  • β-Cyclodextrin as the suitable molecular container for isopulegol enantiomers.
β-Cyclodextrin as the suitable molecular container for isopulegol enantiomers.
Carbohydrate polymers (2013-08-06)
Magdalena Ceborska, Kamila Szwed, Kinga Suwinska
ABSTRACT
Isopulegol, an insoluble in water and highly volatile compound, due to its neuroactive properties is a potentially important agent for medical applications. Formation of "host-guest" molecular complexes with cyclodextrins would lead to the increase of its water solubility and bioavailability. Interactions between native cyclodextrins (α, β and γ) and isopulegol enantiomers were studied in solution proving the formation of inclusion complexes for β- and γ-cyclodextrins. For the more stable complexes with β-cyclodextrin crystal structures were obtained showing the formation of molecular capsules forming molecular container able to accommodate two guest molecules.
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥95.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG