Merck
  • Inhibition of Wnt/β-catenin signaling promotes engraftment of mesenchymal stem cells to repair lung injury.
Inhibition of Wnt/β-catenin signaling promotes engraftment of mesenchymal stem cells to repair lung injury.
Journal of cellular physiology (2013-07-25)
Zhaorui Sun, Xuemin Gong, Huiming Zhu, Cong Wang, Xiaomeng Xu, Di Cui, Weiping Qian, Xiaodong Han
ABSTRACT
We sought to explore the treatment effects and the repair mechanisms of bone marrow derived mesenchymal stem cells (MSCs) during HCl-induced acute lung injury (ALI). MSCs were delivered through the tail veins of rats 24 h after intranasal instillation of HCl. The results showed that MSCs did not ameliorate the histopathologic changes of ALI and pulmonary fibrosis. We found that the activated Wnt/β-catenin signaling may regulate the differentiation of MSCs and is associated with lung fibroblasts activation, pulmonary fibrosis and tissue repair process in ALI rats. Immunofluorescence and histology analysis indicated that activated canonical Wnt/β-catenin signaling induced most MSCs to differentiate into myofibroblasts or fibroblasts in vivo. However, inhibition of Wnt/β-catenin signaling by Dickkopf-1 (DKK1) promotes epithelial differentiation of MSCs induced by native alveolar epithelial cells which are beneficial to repair the injured lung epithelium. Inhibition of Wnt/β-catenin signaling after MSCs transplantation ameliorated pulmonary fibrosis and improved pulmonary function which attenuated the lung injury. In vitro study, activation of the Wnt/β-catenin signaling stimulated MSCs to express myofibroblasts markers, which was attenuated by DKK1. Furthermore, Wnt3α activated Wnt/β-catenin signaling in lung fibroblasts to enhance the expression of collagen I, vimentin and α-smooth muscle actin, but DKK1 attenuated these proteins expression. These findings demonstrated that canonical Wnt/β-catenin signaling plays a key role in regulating differentiation of MSCs in vivo or in vitro and the pathogenesis of fibrotic diseases. Our study suggested that inhibition of abnormal activated Wnt/β-catenin signaling would promote MSCs epithelial differentiation to repair lung injury and reduce pulmonary fibrosis.
Product Description
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid