Merck
  • Biochemical biomarker responses of green-lipped mussel, Perna canaliculus, to acute and subchronic waterborne cadmium toxicity.
Biochemical biomarker responses of green-lipped mussel, Perna canaliculus, to acute and subchronic waterborne cadmium toxicity.
Aquatic toxicology (Amsterdam, Netherlands) (2013-07-24)
Rathishri Chandurvelan, Islay D Marsden, Sally Gaw, Chris N Glover
ABSTRACT
The biochemical responses of the green-lipped mussel, Perna canaliculus, to waterborne cadmium (Cd) were investigated in order to delineate toxic mechanisms, and the impacts of exposure dose and duration, of this important toxicant in a potential sentinel species. Mussels were exposed for either 96 h (acute: 0, 2000, 4000 μgL(-1) Cd) or for 28 d (subchronic: 0, 200, 2000 μgL(-1) Cd), and the digestive gland, gill and haemolymph were examined for impacts. Biochemical responses measured included those associated with metal detoxification (metallothionein-like protein; MTLP), oxidative stress (catalase, lipid peroxidation), cellular homeostasis (alkaline phosphatase, Na(+), K(+)-ATPase; NKA), and energy utilisation (glycogen, haemolymph protein). Following acute exposure, digestive gland glycogen and gill NKA activity were significantly altered by Cd exposure relative to levels in mussels exposed to Cd-free seawater. Subchronic Cd exposure resulted in a significant increase in MTLP levels in both the gill and the digestive gland. This increase was correlated strongly with the levels of Cd accumulation measured in these tissues (R=0.957 for gill, 0.964 for digestive gland). Catalase activity followed a similar pattern, although the correlation with tissue Cd accumulation was not as strong (R=0.907 for gill, 0.708 for digestive gland) as that for MTLP. Lipid peroxidation increased in the digestive gland at Days 7 and 14 at both subchronic Cd levels tested, but this effect had largely dissipated by Days 21 and 28 (with the exception of the 2,000 μgL(-1) group at Day 28). Alkaline phosphatase activity decreased significantly with Cd exposure in both tissues. This effect was observed at both tested concentrations in the gill, but only at the highest concentration for digestive gland. A decrease in digestive gland glycogen levels was observed in Cd-exposed mussels (Days 14 and 21 at 2,000 μgL(-1)), while haemolymph protein levels increased as a result of subchronic Cd exposure. These findings indicated that biochemical responses in Cd-exposed mussels were tissue-specific, dose- and time-dependent, with duration of exposure being the predominant effect. This study shows that biochemical changes in Cd-exposed green-lipped mussels can be linked to tissue metal accumulation and are consistent with previously reported physiological effects. It also suggests that green-lipped mussels are amenable to a multiple biomarker approach and may be of use as a bioindicator species for monitoring coastal metal pollution.
Sigma-Aldrich
Glycogen from oyster
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)