Merck
  • Comparative study of deoxynivalenol, 3-acetyldeoxynivalenol, and 15-acetyldeoxynivalenol on intestinal transport and IL-8 secretion in the human cell line Caco-2.
Comparative study of deoxynivalenol, 3-acetyldeoxynivalenol, and 15-acetyldeoxynivalenol on intestinal transport and IL-8 secretion in the human cell line Caco-2.
Toxicology in vitro : an international journal published in association with BIBRA (2013-06-25)
Tomoyuki Kadota, Hiroko Furusawa, Satoshi Hirano, Osamu Tajima, Yoichi Kamata, Yoshiko Sugita-Konishi
ABSTRACT
The effects of the trichothecene mycotoxin deoxynivalenol (DON) and its acetylated derivatives, 3-acetyldeoxynivalenol (3ADON) and 15-acetyldeoxynivalenol (15ADON) on human intestinal cell Caco-2 were investigated by the studies of transepithelial transport, gene expression, and cytokine secretion. Permeability across a Caco-2 cell monolayer was evaluated by transport study. Transport rates were ranked as DON, 3ADON<15ADON in apical-basolateral direction. 15ADON showed the highest permeability, induced the highest decrease in transepithelial electrical resistance (TEER), and prompted significant Lucifer Yellow permeability. These results showed that 15ADON affect paracellular barrier function extremely. In addition, gene expressions induced by toxins were screened by DNA microarray for investigating cellular effect on Caco-2 cell. The most remarkable gene induced by DON and 15ADON was inflammatory chemokine IL-8 and thus mRNA expression and secretion of IL-8 were analyzed by PCR and ELISA. Both DON and acetylated DONs could induce mRNA expression and production of IL-8. In particular, ELISA assay showed that the ability to produce IL-8 was ranked as 3ADON<DON<15ADON. Our results indicated that 15ADON caused the highest permeability and highest IL-8 secretion among DON, 3ADON, and 15ADON in human intestinal cell.
Product Description
Supelco
3-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Supelco
3-Acetyldeoxynivalenol
Sigma-Aldrich
3-Acetyldeoxynivalenol, from Fusarium roseum
Sigma-Aldrich
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum
Sigma-Aldrich
Deoxynivalenol
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard