Merck
  • Activation of trans geometry in bifunctional mononuclear platinum complexes by a non-bulky methylamine ligand.
Activation of trans geometry in bifunctional mononuclear platinum complexes by a non-bulky methylamine ligand.
Journal of inorganic biochemistry (2013-06-19)
Michaela Frybortova, Olga Novakova, Jana Stepankova, Vojtech Novohradsky, Dan Gibson, Jana Kasparkova, Viktor Brabec
ABSTRACT
In order to shed light on the mechanism that underlies activity of bifunctional mononuclear Pt(II) analogs of transplatin we examined in the present work a DNA binding mode of the analog of transplatin, namely trans-[Pt(CH3NH2)2Cl2], in which NH3 groups were replaced only by a small, non-bulky methylamine ligand. This choice was made because we were interested to reveal the role of the bulkiness of the amines used to substitute NH3 in transplatin to produce antitumor-active Pt(II) drug. The results indicate that trans-[Pt(CH3NH2)2Cl2] forms a markedly higher amount of more distorting intrastrand cross-links than transplatin which forms in DNA preferentially less distorting and persisting monofunctional adducts. Also importantly, the accumulation of trans-[Pt(CH3NH2)2Cl2] in tumor cells was considerably greater than that of transplatin and cisplatin. In addition, the results of the present work demonstrate that the replacement of ammine groups by the non-bulky methylamine ligand in the molecule of ineffective transplatin results in a radical enhancement of its activity in tumor cell lines including cisplatin-resistant tumor cells. Thus, activation of the trans geometry in bifunctional mononuclear Pt(II) complexes can be also accomplished by replacement of ammine groups in transplatin by non-bulky methylamine ligands so that it is not limited only to the replacement by relatively bulky and stereochemically more demanding amino ligands.
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Methylamine hydrochloride, ≥98%