Merck
  • Age-related changes and nutritional regulation of myosin heavy-chain composition in longissimus dorsi of commercial pigs.
Age-related changes and nutritional regulation of myosin heavy-chain composition in longissimus dorsi of commercial pigs.
Animal : an international journal of animal bioscience (2013-06-15)
X M Men, B Deng, Z W Xu, X Tao, K K Qi
ABSTRACT
The objective of this study is to investigate the age-related changes of and the effects of dietary conjugated linoleic acid (CLA) on muscle-fibre types in commercial pigs. We divided 25 crossbred male pigs into five age groups (7, 30, 60, 100 and 180 days) and 30 finishing pigs into two dietary groups (one fed a CLA-enriched diet and the other fed a control diet for 30 days). We analysed the composition (%) of myosin heavy-chain (MyHC) mRNA according to the absolute copies of each MyHC (I, IIa, IIb and IIx) mRNA, and the activities of succinate dehydrogenase (SDH) and malate dehydrogenase (MDH) in the longissimus muscle. From days 7 to 180, the MyHC I mRNA abundance and SDH and MDH activities presented a decreasing trend, the MyHC IIb mRNA abundance presented a steady trend and the MyHC IIa and IIx mRNA abundances presented an increasing trend. On day 30, MyHC I and IIb mRNA abundances were at their lowest (P < 0.05), and the MyHC IIa and IIx mRNA abundances were at their highest (P < 0.05). In the CLA group, the MyHC I mRNA abundance and the activities of SDH and MDH were improved in the longissimus muscle, whereas pressure loss, drip loss and average back fat depth significantly decreased (P < 0.01) and shear force significantly increased (P < 0.01). Loin eye area, feed conversion rate and meat colour showed some tendency to be improved. These results indicated that more oxidative fibres might convert to glycolytic fibres with increasing age or weight, and that the early developmental stage might be a key stage for this conversion. During the finishing stage, the proportion of oxidative fibres might be increased by dietary CLA supplementation, which may contribute to the water-holding capacity of meat. The results would provide an important basis for the application of muscle-fibre types in the improvement of pork quality.
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Malic Dehydrogenase from bovine heart, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)