Merck
  • Effect of high pressure processing on the gel properties of salt-soluble meat protein containing CaCl2 and κ-carrageenan.
Effect of high pressure processing on the gel properties of salt-soluble meat protein containing CaCl2 and κ-carrageenan.
Meat science (2013-05-07)
Fei Ma, Conggui Chen, Lei Zheng, Cunliu Zhou, Kezhou Cai, Zhuo Han
ABSTRACT
The effects of high pressure processing (HPP) on the water-binding capacity and texture profile (TPA) of salt-soluble meat protein (SSMP) containing 0.2% CaCl2 and 0.6% κ-carrageenan (SSMP-CK) gels were investigated. The results showed that 300-400 MPa improved water-binding capacity and decreased TPA parameters of SSMP-CK gels (P<0.05), while 100 MPa could increase hardness and chewiness of the gels. The thermal transition temperature peak for the myosin head (Tpeak1) of SSMP disappeared on addition of CaCl2 and κ-carrageenan. 300 MPa produced a new peak, and caused a shift of the NH-stretching left peak and amide I and the disappearance of NH-stretching right peak. The destruction of network structure and the weakening of molecular interaction within the pressurized gels could result in the decrease of TPA parameters. Thus gelling properties could be modified by HPP, κ-carrageenan and Ca(2+). It is of interest to develop low-fat and sodium-reduced meat products.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, Vetec, reagent grade, 99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.