Merck
  • Lipid profiles are associated with lesion formation over 24 months in interferon-β treated patients following the first demyelinating event.
Lipid profiles are associated with lesion formation over 24 months in interferon-β treated patients following the first demyelinating event.
Journal of neurology, neurosurgery, and psychiatry (2013-04-19)
Bianca Weinstock-Guttman, Robert Zivadinov, Dana Horakova, Eva Havrdova, Jun Qu, Grace Shyh, Elizabeth Lakota, Kerri O'Connor, Darlene Badgett, Miriam Tamaño-Blanco, Michaela Tyblova, Sara Hussein, Niels Bergsland, Laura Willis, Jan Krasensky, Manuela Vaneckova, Zdenek Seidl, Murali Ramanathan
ABSTRACT
To investigate the associations of serum lipid profile with disease progression in high-risk clinically isolated syndromes (CIS) after the first demyelinating event. High density lipoprotein cholesterol (HDL-C), low density lipoprotein cholesterol (LDL-C) and total cholesterol (TC) were obtained in pretreatment serum from 135 high risk patients with CIS (≥ 2 brain MRI lesions and ≥ 2 oligoclonal bands) enrolled in the Observational Study of Early Interferon β-1a Treatment in High Risk Subjects after CIS study (SET study), which prospectively evaluated the effect of intramuscular interferon β-1a treatment following the first demyelinating event. Thyroid stimulating hormone, free thyroxine, 25-hydroxy vitamin D3, active smoking status and body mass index were also obtained. Clinical and MRI assessments were obtained within 4 months of the initial demyelinating event and at 6, 12 and 24 months. The time to first relapse and number of relapses were not associated with any of the lipid profile variables. Higher LDL-C (p=0.006) and TC (p=0.001) levels were associated with increased cumulative number of new T2 lesions over 2 years. Higher free thyroxine levels were associated with lower cumulative number of contrast-enhancing lesions (p=0.008). Higher TC was associated as a trend with lower baseline whole brain volume (p=0.020). Higher high density lipoprotein was associated with higher deseasonalised 1,25-dihydroxy vitamin D3 (p=0.003) levels and a trend was found for deseasonalised 25-hydroxy vitamin D3 (p=0.014). In early multiple sclerosis, lipid profile variables particularly LDL-C and TC levels are associated with inflammatory MRI activity measures.
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Interferon β-1a, European Pharmacopoeia (EP) Reference Standard