Merck
  • Nanorod-nanosheet hierarchically structured ZnO crystals on zinc foil as flexible photoanodes for dye-sensitized solar cells.
Nanorod-nanosheet hierarchically structured ZnO crystals on zinc foil as flexible photoanodes for dye-sensitized solar cells.
Nanoscale (2013-01-29)
Rui Gao, Jianjun Tian, Zhiqiang Liang, Qifeng Zhang, Liduo Wang, Guozhong Cao
ABSTRACT
In this paper, ZnO nanorod-nanosheet hierarchical structures were fabricated using a facile method on zinc foil and used as flexible photoanodes in dye-sensitized solar cells (DSCs). Compared to nanorods (NRs) obtained by the dissolution-precipitation method, the nanorod-nanosheet (NR-NS) hierarchical structures obtained by a second-step homogeneous precipitation improved the performance of DSCs by increasing photocurrent density significantly. As a result, the power conversion efficiency of the devices based on such a NR-NS hierarchical structure reached up to 2.4% under 100 mW cm(-2) illumination condition. This represents an enhancement by 108% as compared to DSCs based on NR assembled nanoflowers, for which the efficiency was 1.1%. The enhancement of the photocurrent was due mainly to the much larger specific surface area and resulting dye-loading amount. The electron transport properties in this structure were also investigated by means of electrochemical impedance spectroscopy (EIS). Furthermore, the formation mechanism of the NR-NS hierarchical structures are discussed.
Product Description
Zinc, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.015mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 300x300mm, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.015mm, 99.9%
Zinc, foil, 15mm disks, thickness 0.01mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 1.5mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%