- Selective synthesis of nickel oxide nanowires and length effect on their electrochemical properties.
Selective synthesis of nickel oxide nanowires and length effect on their electrochemical properties.
Nanoscale (2010-07-22)
Huan Pang, Qingyi Lu, Yizhou Zhang, Yecheng Li, Feng Gao
PMID20648289
ABSTRACT
Nickel oxide nanostructures with different lengths have been easily synthesized by calcining nickel foil after soaking in LiOH solution of different concentrations. The measurements of electrochemical properties of these NiO nanostructures show that different lengths of NiO nanostructures would have different electrochemical properties and the NiO nanowires with the longest length have the largest specific capacitance of 180.00 F g(-1).
Product Description
Nickel, foil, light tested, 50x50mm, thickness 0.0075mm, 99.95%
Nickel, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 0.5m coil, thickness 1.0mm, annealed, 99.98%
Nickel, foil, 4mm disks, thickness 0.05mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, 0.5m coil, thickness 0.0025mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.01mm, 99.95%
Nickel, foil, not light tested, 50x50mm, thickness 0.004mm, hard, 99%
Nickel, foil, 10m coil, thickness 0.3mm, coil width 1mm, as rolled, 99%
Nickel, foil, 10m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.20mm, annealed, 99%
Nickel, foil, 50x50mm, thickness 0.50mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 2.0mm, as rolled, 99.99+%
Nickel, foil, 25x25mm, thickness 0.15mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.01mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.008mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.003mm, 99.95%
Nickel, foil, 4mm disks, thickness 0.0075mm, 99.9%
Nickel, foil, 10m coil, thickness 0.20mm, coil width 1mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.015mm, 99.9%
Nickel, foil, 10mm disks, thickness 0.0075mm, 99.95%
Nickel, foil, 2m coil, thickness 0.30mm, as rolled, 99.98%
Nickel, foil, 25x25mm, thickness 0.3mm, as rolled, 99.999%
Nickel, foil, 4mm disks, thickness 0.0075mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.025mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.1mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.01mm, 99.95%
Nickel, foil, 10m coil, thickness 0.125mm, coil width 1mm, as rolled, 99%
Nickel, foil, 4mm disks, thickness 0.05mm, as rolled, 99.98%