Merck
  • Calorimetric and computational study of 3-buten-1-ol and 3-butyn-1-ol. Estimation of the enthalpies of formation of 1-alkenols and 1-alkynols.
Calorimetric and computational study of 3-buten-1-ol and 3-butyn-1-ol. Estimation of the enthalpies of formation of 1-alkenols and 1-alkynols.
The journal of physical chemistry. A (2006-07-13)
Ederley Vélez, Jairo Quijano, Jair Gaviria, María Victoria Roux, Pilar Jiménez, Manuel Temprado, Gloria Martín-Valcárcel, Juan Pérez-Parajón, Rafael Notario
ABSTRACT
The enthalpies of combustion and vaporization of 3-buten-1-ol and 3-butyn-1-ol have been measured by static bomb combustion calorimetry and correlation gas chromatography techniques, respectively, and the gas-phase enthalpies of formation, Delta(f)H degrees (m)(g), have been determined, the values being -147.3 +/- 1.8 and 16.7 +/- 1.6 kJ mol(-1), for 3-buten-1-ol and 3-butyn-1-ol, respectively. High level calculations at the G2 and G3 levels have also been carried out. Relationships between the enthalpies of formation of 1-alkanols, 1-alkenols and 1-alkynols and with the corresponding hydrocarbons have been discussed. From the calculated contributions to Delta(f)H degrees (m)(g) for the substitutions of CH(3) by CH(2)OH, CH(3)CH(2) by CH(2)=CH and CH(3)CH(2) by CH triple bond C, we have estimated the Delta(f)H degrees (m)(g) values for 3-buten-1-ol and 3-butyn-1-ol, in excellent agreement with the experimental ones. Delta(f)H degrees (m)(g) values for 1-alkenols and 1-alkynols up to 10 carbon atoms have also been estimated.
Sigma-Aldrich
3-Butyn-1-ol, 97%
Sigma-Aldrich
3-Buten-1-ol, 96%