Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • An improved sample preparation method for the sensitive detection of peptides by MALDI-MS.
An improved sample preparation method for the sensitive detection of peptides by MALDI-MS.
Journal of mass spectrometry : JMS (2013-11-22)
Yusaku Hioki, Hiroki Kuyama, Chikako Hamana, Kohei Takeyama, Koichi Tanaka
ABSTRACT
We describe here an optimization study of the sample preparation conditions for sensitive detection of peptides by matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS). Among many factors in the conditions, we varied the percent acetonitrile in the peptide solution, the percent acetonitrile in the matrix solution and the α-cyano-4-hydroxycinnamic acid (CHCA) concentration in the matrix solution. CHCA was chosen because it is the most frequently used matrix for analyzing peptides. The well-established dried-droplet method was employed for sample deposition. The examined range of the concentration of CHCA was from 0.01 to 10 mg/ml, and the MeCN content of the solvent for matrix/analyte was 10% to 50%. The indicator for the detection sensitivity was the S/N ratio of the peaks of peptides used. Highly increased sensitivity (100- to 1000-fold) was observed for the optimal CHCA concentration of 0.1 mg/ml in 20% MeCN/0.1% aq. trifluoroacetic acid (TFA), as compared with the conventional concentration (10 mg/ml) in 50% MeCN/0.1% aq. TFA. For example, the limit of detection of human ACTH 18-39 was 10 amol/well for the optimal condition but 10 fmol/well for the conventional condition. The optimal condition (0.1 mg/ml CHCA in 20% MeCN/0.1% aq. TFA) was verified with five model peptides and provided significant improvement in sensitivity (by two to three orders of magnitude) compared with the conventional conditions. Optimizing the CHCA concentration and solvent composition significantly improved the detection sensitivity in the analysis of peptides by MALDI-MS.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%