- Miniaturizable homogenous time-resolved fluorescence assay for carboxypeptidase B activity.
Miniaturizable homogenous time-resolved fluorescence assay for carboxypeptidase B activity.
Analytical biochemistry (2003-05-06)
Marc Ferrer, Paul Zuck, Garrett Kolodin, Shi Shan Mao, Richard R Peltier, Carolyn Bailey, Stephen J Gardell, Berta Strulovici, James Inglese
PMID12729605
ABSTRACT
An epitope-unmasking, homogeneous time-resolved fluorescence (HTRF) assay has been developed for measuring carboxypeptidase B (CPB) activity in a miniaturized high-throughput screening format. The enzyme substrate (biotin-RYRGLMVGGVVR-OH) is cleaved by CPB at the C terminus, causing release of the C-terminal Arg residue. The product (biotin-RYRGLMVGGVV-OH) is recognized specifically by a monoclonal antibody (G2-10) which is labeled with Eu(3+)-cryptate ([Eu(3+)]G2-10 mAb), and the complex is detected by fluorescence resonance energy transfer using streptavidin labeled with allophycocyanin ([XL665]SA). The CPB HTRF assay is readily adapted from 96- to 1536-well format as a robust (Z(')>0.5) assay for high-throughput screening.