Merck
  • Highly efficient synthesis of N-substituted isoindolinones and phthalazinones using Pt nanowires as catalysts.
Highly efficient synthesis of N-substituted isoindolinones and phthalazinones using Pt nanowires as catalysts.
Organic letters (2012-03-17)
Linyan Shi, Lei Hu, Jiaqing Wang, Xueqin Cao, Hongwei Gu
ABSTRACT
A series of N-substituted isoindolinones have been successfully synthesized through the reductive C-N coupling and intramolecular amidation of 2-carboxybenzaldehyde and amines. This one-pot synthesis gives excellent yields using ultrathin Pt nanowires as catalysts under 1 bar of hydrogen. These unsupported catalysts can also be used for the synthesis of phthalazinones in high yield when hydrazine or phenyl hydrazine is used instead of amines.
Sigma-Aldrich
2-Carboxybenzaldehyde, 97%