Merck
  • The occurrence of 2-hydroxyisovaleric acid in patients with lactic acidosis and ketoacidosis.
The occurrence of 2-hydroxyisovaleric acid in patients with lactic acidosis and ketoacidosis.
Clinica chimica acta; international journal of clinical chemistry (1977-08-01)
S Landaas, C Jakobs
PMID884872
ABSTRACT
Examination of urine samples by capillary column gas chromatography and mass spectrometry showed the presence of 2-hydroxyisovaleric acid together with trace amounts of the three branched-chain 2-keto acids in five patients with lactic and ketoacidosis. None of these compounds was detected in urine from healthy controls.
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%