Merck
  • Cystinylglycine in plasma: diagnostic relevance for pyroglutamic acidemia, homocystinuria, and phenylketonuria.
Cystinylglycine in plasma: diagnostic relevance for pyroglutamic acidemia, homocystinuria, and phenylketonuria.
Clinica chimica acta; international journal of clinical chemistry (1981-11-25)
T L Perry, S Hansen
ABSTRACT
Cystinylglycine, recently identified as a normal small peptide in human plasma, has diagnostic importance for several genetically determined disorders. We found cystinylglycine absent from the plasma of a patient with pyroglutamic acidemia, and the peptide was either absent or greatly reduced in plasma from patients with homosyctinuria. In the latter disorder, a different small peptide replaced cystinylglycine. It was identified as the mixed disulfide of homocysteine and cysteinylglycine. The mean plasma concentration of cystinylglycine was 13.6 +/- 3.6 mumol/l in adult control subjects, and concentrations of the mixed disulfide of homocysteine and cysteinylglycine varied between 2 and 10 mumol/l in the plasma of homocystinuric patients. Failure to separate cystinylglycine from phenylalanine with many rapid amino acid analyzer systems can lead to a misclassification of persons as heterozygotes for the phenylketonuria gene when heterozygosity testing is based on the phenylalanine/tyrosine molar ratio in fasting plasma.
Product Description
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Sigma-Aldrich
2-Hydroxyphenylacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DL-Mandelic acid, analytical standard
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%
Sigma-Aldrich
Phenylpyruvic acid, 98%
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%