Merck
  • Low CSF GABA concentration in children with febrile convulsions, untreated epilepsy, and meningitis.
Low CSF GABA concentration in children with febrile convulsions, untreated epilepsy, and meningitis.
Journal of neurology (1983-01-01)
D Rating, H Siemes, W Löscher
ABSTRACT
In 14 children with epilepsy, 51 with febrile convulsions and 22 with meningitis gamma-aminobutyric acid (GABA) concentrations in lumbar CSF were determined. While the mean for CSF GABA concentrations for all epileptic children was unchanged [144 (range: 73-285) pmol/ml; controls: 148 (range: 90-243) pmol/ml] extraordinarily high GABA levels were found in the CSF of two children on valproate (525 and 557 pmol/ml) and remarkably low GABA concentrations in hitherto untreated epileptic children [109 (range: 67-176) pmol/ml]. Children with febrile convulsions [103 (range: 63-170) pmol/ml] and acute meningitis [105 (range: 65-171) pmol/ml] had significantly decreased CSF GABA concentrations (P less than 0.001 and P less than 0.02 compared with controls). The data indicate that valproate intake increases dramatically the GABA concentrations in the CSF of epileptic children. Furthermore, the study supports the concept that low GABAergic activity within the CNS may be one cause for an increased seizure frequency.
Product Description
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(R)-3-Hydroxybutyric acid, ≥98.0% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Citrulline, ≥98% (TLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Isoleucine