Merck
  • The occurrence of gamma-glutamylphenylalanine in the urine of newborn phenylketonurics.
The occurrence of gamma-glutamylphenylalanine in the urine of newborn phenylketonurics.
Clinica chimica acta; international journal of clinical chemistry (1979-06-15)
H Peck, R J Pollitt
PMID466810
ABSTRACT
The urine of untreated phenylketonurics in the first weeks of life contains gamma-glutamylphenylalanine (0.07--0.69 mmol/g creatinine, 12 samples) visible on the normal 2-dimensional electrophoreto-chromatogram. This compound is less prominent or absent when urine from older untreated phenylketonurics is examined and is not seen in normal urine.
Product Description
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Sigma-Aldrich
2-Hydroxyphenylacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%
Sigma-Aldrich
Phenylpyruvic acid, 98%
Supelco
DL-Mandelic acid, analytical standard